missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ ActiCHO™ SM Powdered Media

Catalog No. SH3102906
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3102906 1 L
SH3102901 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3102906 Supplier Cytiva Supplier No. SH31029.06
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrolysate-free and animal-derived component-free (ADCF)

ActiCHO SM powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.

  • Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides or growth factors in ActiCHO SM medium.

Specifications

Form Powder
Sterility Non-sterile
With Additives Poloxamer 188
Without Additives No Glutamine, No Insulin
Quantity 1 L
Content And Storage 2 to 8°C, away from light and moisture