missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ 100x Vitamin Solution

Catalog No. SH3059901
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3059901 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3059901 Supplier Cytiva Supplier No. SH30599.01

Specifications

Certifications/Compliance cGMP and ISO 91:2 Guidelines
Description MEM Vitamin Solution, 100x
Concentration 1 X
Sterilization Method 0.1 μm Sterile-filtered
Without Additives No Serum, No Protein
Endotoxin Level =1.0EU/mL
Quantity 100 mL