Learn More
Rotronic™ Humidity standards
For on-site calibration and adjustment of ROTRONIC probes with handheld instrument
$635.86 - $638.31
Specifications
Certificate of Analysis SCS traceable calibration certificate, textile pad, calibration instruction
Technique On-site Calibration and Adjustment of Rotronic Probe
For Use With (Equipment) Rotronic Instrument
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
1165013 DFS Item
View Documents
Rotronic™
EA10SCS
Humidity Standard (Certified)
Case of 5 Each for $638.31
Only null left
11-650-6
View Documents
Rotronic™
5010EA80SCS1
Humidity Standard
Pack of 5 Each for $635.86
Only null left
Description
Description
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
  • Ampoules contain unsaturated salt solutions
  • Simple and safe on-site calibration method
  • Unlimited storage life
Specifications
Specifications
SCS traceable calibration certificate, textile pad, calibration instruction
On-site Calibration and Adjustment of Rotronic Probe
Rotronic Instrument
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'