missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Humicade Chimeric Recombinant Mouse Monoclonal Antibody (CDP 571 (Humicade))
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551587
Description
The antibody binds to human TNF-alpha with a Kd of 0.087 nM.
The structure of Humicade is a monoclonal antibody that is highly similar to the reference medicine, Humira (adalimumab). It is composed of human immunoglobulin G1 (IgG1) antibodies that are produced using recombinant DNA technology. The function of Humicade is to treat certain autoimmune conditions by blocking the action of tumor necrosis factor-alpha (TNF-alpha), a protein that contributes to inflammation. The structure of Humicade allows it to specifically target TNF-alpha and block its activity, reducing inflammation and alleviating symptoms in certain autoimmune conditions.Specifications
| Humicade Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Human TNF-alpha. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Neutralization | |
| CDP 571 (Humicade) | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |