missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF85 (aa 62-87) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100034
Description
Highest antigen sequence indentity to the following orthologs: Mouse (35%), Rat (35%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60818 (PA5-60818. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
| Q03923 | |
| Blocking Assay, Control | |
| 7639 | |
| 100 μL | |
| HPF4; HTF1; Zinc finger protein 85; zinc finger protein 85 (HPF4, HTF1); Zinc finger protein HPF4; Zinc finger protein HTF1; ZNF85 | |
| ZNF85 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human ZNF85 (aa 62-87) Control Fragment | |
| RUO | |
| ZNF85 | |
| Unconjugated | |
| Recombinant | |
| AWSMKRHEIMVAKPTESYSVTQSGMQ | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |