missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF664 (aa 4-27) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP106156
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65482 (PA5-65482. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in transcriptional regulation.Specifications
| Q8N3J9 | |
| Blocking Assay, Control | |
| 144348 | |
| 100 μL | |
| AW060232; D930038J03Rik; ZFOC1; Zfp664; zinc finger Organ of Corti 1; zinc finger protein; zinc finger protein 176; Zinc finger protein 664; zinc finger protein from organ of Corti; zinc finger protein ZFOC1; ZNF176; ZNF196; Znf664 | |
| ZNF664 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human ZNF664 (aa 4-27) Control Fragment | |
| RUO | |
| ZNF664 | |
| Unconjugated | |
| Recombinant | |
| KCPMCREFFSERADLFMHQKIHTA | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |