missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF628 (aa 810-888) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP93642
Description
Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60716 (PA5-60716. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Zinc finger proteins (ZNFs), which bind nucleic acids, perform many key functions, the most important of which is regulating transcription.Specifications
| Q5EBL2 | |
| Blocking Assay, Control | |
| 89887 | |
| 100 μL | |
| Zec; Zfp628; Zinc finger protein 628; zinc finger protein expressed in embryonal cells and certain adult organs; zinc finger protein Zec; Znf628 | |
| ZNF628 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human ZNF628 (aa 810-888) Control Fragment | |
| RUO | |
| ZNF628 | |
| Unconjugated | |
| Recombinant | |
| AGPQEMSGVQLQPLRPAPEVTTVQLQPAQEVTTVQLQPAQEVTTVQLQPAQEVTTVQLQPVAGQLSNSSGGAVATEAPN | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |