missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF619 (aa 112-207) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP99936

Catalog No. RP99936

Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (28%), Rat (28%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60921 (PA5-60921. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in transcriptional regulation.
Specifications

Specifications

Q8N2I2
Blocking Assay, Control
285267
100 μL
Zinc finger protein 619; ZNF619
ZNF619
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human ZNF619 (aa 112-207) Control Fragment
RUO
ZNF619
Unconjugated
Recombinant
GLLMDVPQHPDFKDRLEKSQLHDTGNKTKIGDCTDLTVQDHESSTTEREEIARKLEESSVSTHLITKQGFAKEQVFYKCGECGSYYNPHSDFHLHQ
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions