missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF618 (aa 746-844) Control Fragment Recombinant Protein

Catalog No. RP93021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93021 Supplier Invitrogen™ Supplier No. RP93021

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54698 (PA5-54698. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in transcriptional regulation.

Specifications

Accession Number Q5T7W0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 114991
Name Human ZNF618 (aa 746-844) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias FP13169; KIAA1952; NEDD10; neural precursor cell expressed, developmentally down-regulated 10; zinc finger protein 618; ZNF618
Common Name ZNF618
Gene Symbol ZNF618
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VIELSNESQPTLQLVLPTYVRLEKLFTAKANDAGTVSKLCHLFLEALKENFKVHPAHKVAMILDPQQKLRPVPPYQHEEIIGKVCELINEVKESWAEEA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less