missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF442 (aa 6-33) Control Fragment Recombinant Protein

Catalog No. RP100542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100542 Supplier Invitrogen™ Supplier No. RP100542
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (43%), Rat (43%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61084 (PA5-61084. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in transcriptional regulation.

Specifications

Accession Number Q9H7R0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79973
Name Human ZNF442 (aa 6-33) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias LOC100909622; Zinc finger protein 442; zinc finger protein 442-like; ZNF442
Common Name ZNF442
Gene Symbol ZNF442
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GEDRSDLFLPDSQTNEERKQYDSVAFED
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less