missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZBTB14 (aa 81-173) Control Fragment Recombinant Protein

Catalog No. RP91702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91702 Supplier Invitrogen™ Supplier No. RP91702

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62236. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transcriptional activator of the dopamine transporter (DAT), binding it's promoter at the consensus sequence 5'-CCTGCACAGTTCACGGA-3'. Binds to 5'-d(GCC)(n)-3' trinucleotide repeats in promoter regions and acts as a repressor of the FMR1 gene. Transcriptional repressor of MYC and thymidine kinase promoters.

Specifications

Accession Number O43829
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 7541
Name Human ZBTB14 (aa 81-173) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias b2b1982Clo; Zbtb14; ZF5; ZFP161; zfp-161; ZFP161 zinc finger protein; Zfp5; Zfp-5; zinc finger and BTB domain containing 14; zinc finger and BTB domain-containing protein 14; Zinc finger protein 161; zinc finger protein 161 homolog; Zinc finger protein 478; zinc finger protein 5; Zinc finger protein 5 homolog; zinc finger protein homologous to Zfp161 in mouse; ZNF478
Common Name ZBTB14
Gene Symbol ZBTB14
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LRSDIFEEVLNYMYTAKISVKKEDVNLMMSSGQILGIRFLDKLCSQKRDVSSPDENNGQSKSKYCLKINRPIGDAADTQDDDVEEIGDQDDSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less