missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human XPOT (aa 126-212) Control Fragment Recombinant Protein

Catalog No. RP106169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106169 Supplier Invitrogen™ Supplier No. RP106169
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65492 (PA5-65492. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.

Specifications

Accession Number O43592
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11260
Name Human XPOT (aa 126-212) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110004L07Rik; 3110065H13Rik; AI452076; C79645; exportin for tRNA; exportin(tRNA); exportin, tRNA; exportin, tRNA (nuclear export receptor for tRNAs); exportin-T; tRNA exportin; XPO3; XPOT
Common Name XPOT
Gene Symbol XPOT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FDILSVVDLNPRGVDLYLRILMAIDSELVDRDVVHTSEEARRNTLIKDTMREQCIPNLVESWYQILQNYQFTNSEVTCQCLEVVGAY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less