missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Wnt-5b (NP_110402.2) VersaClone cDNA

Specifications
Specifications
| Gene | Wnt-5b |
| Accession Number | NP_110402.2 |
| Gene ID (Entrez) | 81029 |
| Gene Alias | MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, Wnt5b, WNT-5B protein |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |