missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Wnt-4 ELISA Kit

Catalog No. EH485RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH485RBX5 5 x 96 Tests
EH485RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH485RBX5 Supplier Invitrogen™ Supplier No. EH485RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

ELISA

Human Wnt-4 quantitates human Wnt-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Wnt-4.

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family, and is the first signaling molecule shown to influence the sex-determination cascade. It encodes a protein which shows 98% amino acid identity to the Wnt4 protein of mouse and rat. This gene and a nuclear receptor known to antagonize the testis-determining factor play a concerted role in both the control of female development and the prevention of testes formation. This gene and another two family members, WNT2 and WNT7B, may be associated with abnormal proliferation in breast tissue. Mutations in this gene can result in Rokitansky-Kuster-Hauser syndrome and in SERKAL syndrome.

Specifications

Accession Number P56705
Assay Range 1.22-300 ng/mL
Assay Sensitivity 1.22 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias protein Wnt-4; SERKAL; signal molecule; UNQ426/PRO864; wingless-related MMTV integration site 4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4; Wnt family member 4; WNT4; Wnt-4protein Wnt-4; SERKAL; signal molecule; UNQ426/PRO864; wingless-related MMTV integration site 4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4; Wnt family member 4; WNT4; Wnt-4;
Gene ID (Entrez) 54361
Gene Symbol Wnt4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target WNT4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less