missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Wnt-10a (NP_079492.2) VersaClone cDNA
Specifications
Specifications
| Gene | Wnt-10a |
| Accession Number | NP_079492.2 |
| Gene ID (Entrez) | 80326 |
| Gene Alias | FLJ14301, protein Wnt-10a, SSPS, wingless-type MMTV integration site family, member 10A, Wnt10a |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |