missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VWF Native Protein

Catalog No. PIRP43132
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43132 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43132 Supplier Invitrogen™ Supplier No. RP43132
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43132 contains native human von Willebrand Factor protein. RP-43132 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.

Specifications

Accession Number P04275
Concentration 0.25 mg/mL
For Use With (Application) Neutralization
Formulation 25mM sodium citrate with 100mM glycine, 100mM NaCl and no preservative; pH 6.8
Gene ID (Entrez) 7450
Molecular Weight (g/mol) 260 kDa
Name Human VWF
pH Range 6.8
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312
Common Name VWF
Gene Symbol VWF
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -80° C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less