missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human VPS52 (aa 581-677) Control Fragment Recombinant Protein

Catalog No. RP99253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99253 Supplier Invitrogen™ Supplier No. RP99253

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61662 (PA5-61662. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that is similar to the yeast suppressor of actin mutations 2 gene. The yeast protein forms a subunit of the tetrameric Golgi-associated retrograde protein complex that is involved in vesicle trafficking from both early and late endosomes, back to the trans-Golgi network. This gene is located on chromosome 6 in a head-to-head orientation with the gene encoding ribosomal protein S18.

Specifications

Accession Number Q8N1B4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6293
Name Human VPS52 (aa 581-677) Control Fragment
Quantity 100 μL
Gene Alias Are1; D130068D18; D430041K17Rik; dJ1033B10.5; DKFZp547I194; fb94c11; LOW QUALITY PROTEIN: vacuolar protein sorting-associated protein 52 homolog; RP5-1033B10; SAC2; SAC2 suppressor of actin mutations 2-like protein; SACM2L; tclw5; vacuolar protein sorting 52 (yeast); vacuolar protein sorting 52 homolog; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; vacuolar protein sorting-associated protein 52-like protein; Vacuolar protein sorting-associated protein 52-like protein-like protein; Vps52; VPS52 GARP complex subunit; VPS52 subunit of GARP complex; VPS52, GARP complex subunit; wu:fb94c11
Common Name VPS52
Gene Symbol VPS52
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DDSKEVESFQQLLNARTQEFIEELLSPPFGGLVAFVKEAEALIERGQAERLRGEEARVTQLIRGFGSSWKSSVESLSQDVMRSFTNFRNGTSIIQGA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less