missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Vitronectin Native Protein

Catalog No. PIRP43152
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43152 Supplier Invitrogen™ Supplier No. RP43152

missing translation for 'validMainframeMsg'

Native Protein

RP-43152 contains native human Vitronectin protein. RP-43152 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.

Specifications

Accession Number P04004
Concentration 0.9 mg/mL
For Use With (Application) Neutralization
Formulation PBS with no preservative; pH 7.4
Gene ID (Entrez) 7448
Molecular Weight (g/mol) 75 kDa
Name Human Vitronectin
pH Range 7.4
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Common Name Vitronectin
Gene Symbol VTN
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less