missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Vitronectin, Animal-Free Recombinant Protein, PeproTech®

Catalog No. p-7245937
Encompass
Change view
Click to view available options
Quantity:
100 μg
500 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF14009100 100 μg
AF140091MG 1 mg
AF14009500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AF14009100 Supplier Gibco™ Supplier No. AF14009100UG
Only null left

Recombinant Protein

Recombinant Human Vitronectin is a 459 amino acid, single-chain, monomeric protein, which migrates at an apparent molecular weight of 75 kDa by SDS-PAGE under reducing conditions. The calculated molecular weight of Recombinant Human Vitronectin is 52.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.

Specifications

Accession Number P04004
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7448
Molecular Weight (g/mol) 52.2 kDa
Name Human Vitronectin, Animal-Free
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Common Name Vitronectin
Gene Symbol VTN
Biological Activity Recombinant human Vitronectin promotes attachment of hESC and iPSC in serum-free, feeder conditions at 5 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence DQESCKGRCT EGFNVDKKCQ CDELCSYYQS CCTDYTAECK PQVTRGDVFT MPEDEYTVYD DGEEKNNATV HEQVGGPSLT SDLQAQSKGN PEQTPVLKPE EEAPAPEVGA SKPEGIDSRP ETLHPGRPQP PAEEELCSGK PFDAFTDLKN GSLFAFRGQY CYELDEKAVR PGYPKLIRDV WGIEGPIDAA FTRINCQGKT YLFKGSQYWR FEDGVLDPDY PRNISDGFDG IPDNVDAALA LPAHSYSGRE RVYFFKGKQY WEYQFQHQPS QEECEGSSLS AVFEHFAMMQ RDSWEDIFEL LFWGRTSAGT RQPQFISRDW HGVPGQVDAA MAGRIYISGM APRPSLAKKQ RFRHRNRKGY RSQRGHSRGR NQNSRRPSRA TWLSLFSSEE SNLGANNYDD YRMDWLVPAT CEPIQSVFFF SGDKYYRVNL RTRRVDTVDP PYPRSIAQYW LGCPAPGHL
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less