Learn More
Invitrogen™ Human VEGF Receptor 2/KDR ELISA Kit
ELISA
Specifications
| Accession Number | P35968 |
|---|---|
| Assay Range | 78 to 5,000 pg/mL |
| Assay Sensitivity | 7.0 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human VEGF-R2/KDR
Biotin-Conjugate anti-human VEGF-R2/KDR monoclonal antibody
Streptavidin-HRP
Human VEGF-R2/KDR Standard lyophilized, 10ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
5017887
|
Invitrogen™
BMS2019 |
96 Tests |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
5017889
|
Invitrogen™
BMS2019TEN |
10 x 96 Tests |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Human Vascular Endothelial Growth Factor Receptor 2/KDR (Hu VEGFR2) ELISA quantitates Hu VEGFR2 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu VEGFR2. Principle of the method The Human VEGFR2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.Specifications
| P35968 | |
| 7.0 pg/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CD309,FLK1,VEGFR,VEGFR2 | |
| 5.3 | |
| HRP | |
| Plasma 20 μL, Serum 20 μL, Supernatant 20 μL | |
| KDR, FLK1, Kinase insert domain receptor, CD309 | |
| 1 hrs 20 mins |
| 78 to 5,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 3791 | |
| 9.8% | |
| Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| FLK1 (VEGF Receptor 2) | |
| 3 hrs 30 mins |