Learn More
Invitrogen™ Human VEGF-165 ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier: Invitrogen™ 900K10K
Description
The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.Specifications
| P15692 | |
| Lot-specific | |
| ELISA | |
| 7422 | |
| Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
| 10 x 96 Tests | |
| Refer to product documentation for component specific storage temperature. | |
| 1 hrs 45 mins |
| Lot-specific | |
| HRP | |
| Colorimetric Microplate Reader | |
| Vegfa | |
| HRP | |
| RUO | |
| VEGF-165 | |
| 24 hrs 30 mins |