missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human UTP23 (aa 85-166) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60705 (PA5-60705. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9BRU9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 84294 |
| Name | Human UTP23 (aa 85-166) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700010I21Rik; AI662478; BB238373; C8orf53; D530033C11Rik; RGD1307179; rRNA-processing protein UTP23 homolog; UTP23; UTP23, small subunit (SSU) processome component, homolog; UTP23, small subunit (SSU) processome component, homolog (yeast); UTP23, small subunit processome component |
| Common Name | UTP23 |
| Show More |