missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human USP36 (aa 971-1115) Control Fragment Recombinant Protein

Catalog No. RP106421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106421 Supplier Invitrogen™ Supplier No. RP106421

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65745 (PA5-65745. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 'Lys-4' trimethylation (H3K4).

Specifications

Accession Number Q9P275
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57602
Name Human USP36 (aa 971-1115) Control Fragment
Quantity 100 μL
Gene Alias 2700002L06Rik; deubiquitinating enzyme 36; DUB1; Kiaa1453; mKIAA1453; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific peptidase 36; ubiquitin specific protease 36; ubiquitin thioesterase 36; ubiquitin thiolesterase 36; ubiquitin-specific-processing protease 36; Usp36
Common Name USP36
Gene Symbol USP36
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EDGHLKCPRSAKPQDAVVPESSSCAPSANGWCPGDRMGLSQAPPVSWNGERESDVVQELLKYSSDKAYGRKVLTWDGKMSAVSQDAIEDSRQARTETVVDDWDEEFDRGKEKKIKKFKREKRRNFNAFQKLQTRRNFWSVTHPAK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less