missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | bA350J20.7; LETAL; Lymphocyte effector toxicity activation ligand; MGC125309; N2DL4; N2DL-4; NKG2D ligand 4; NKG2DL4; Rae-1 epsilon; RAE1 like transcript 4; RAE1epsilon; RAE-1-epsilon; RAE-1-like transcript 4; Raet1e; RAET1E2; Retinoic acid early transcript 1 E; retinoic acid early-inducible protein 1-epsilon; RL-4; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4; ULBP4MGC125308; UNQ1867/PRO4303 |
|---|---|
| Assay Range | 0.43 to 100 ng/mL |
| Assay Sensitivity | 0.43 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH479RBX5
|
Invitrogen™
EH479RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH479RB
|
Invitrogen™
EH479RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human ULBP-4/RAET1E quantitates human ULBP-4/RAET1E in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ULBP-4/RAET1E.
This gene belong to the RAET1 family, which consists of major histocompatibility complex (MHC) class I-related genes located in a cluster on chromosome 6q24.2-q25.3. This and RAET1G protein differ from other RAET1 proteins in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. This protein functions as a ligand for NKG2D receptor, which is expressed on the surface of several types of immune cells, and is involved in innate and adaptive immune responses. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Aug 2011].Specifications
| bA350J20.7; LETAL; Lymphocyte effector toxicity activation ligand; MGC125309; N2DL4; N2DL-4; NKG2D ligand 4; NKG2DL4; Rae-1 epsilon; RAE1 like transcript 4; RAE1epsilon; RAE-1-epsilon; RAE-1-like transcript 4; Raet1e; RAET1E2; Retinoic acid early transcript 1 E; retinoic acid early-inducible protein 1-epsilon; RL-4; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4; ULBP4MGC125308; UNQ1867/PRO4303 | |
| 0.43 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| RAET1E | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| N2DL4 | |
| 4 hrs 45 mins |
| 0.43 to 100 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 135250 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |