missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TXNDC6 (aa 169-248) Control Fragment Recombinant Protein

Catalog No. RP104035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104035 Supplier Invitrogen™ Supplier No. RP104035

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83483 (PA5-83483. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be a regulator of microtubule physiology.

Specifications

Accession Number Q86XW9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 347736
Name Human TXNDC6 (aa 169-248) Control Fragment
Quantity 100 μL
Gene Alias MGC129586; NM23-H9; NME gene family member 9; NME/NM23 family member 9; NME9; Thioredoxin domain-containing protein 6; thioredoxin-like 2; thioredoxin-like protein 2; TXL2; Txl-2; TXNDC6
Common Name TXNDC6
Gene Symbol NME9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AVAHGKTDEIIMKIQEAGFEILTNEERTMTEAEVRLFYQHKAGEEAFEKLVHHMCSGPSHLLILTRTEGFEDVVTTWRTV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less