missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TUBA1A (aa 26-53) Control Fragment Recombinant Protein

Catalog No. RP104318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104318 Supplier Invitrogen™ Supplier No. RP104318
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58711 (PA5-58711. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.

Specifications

Accession Number Q71U36
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7846
Name Human TUBA1A (aa 26-53) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias alpha 1 A antibody; alpha 3 antibody; alpha-tubulin; alpha-tubulin 1; Alpha-tubulin 3; alpha-tubulin 3 antibody; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; B-ALPHA-1 antibody; brain-specific antibody; Detyrosinated tubulin alpha-1 A chain; FLJ25113 antibody; FLJ30169; H2 ALPHA; H2ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub1 antibody; hum-a-tub2; hum-a-tub2 antibody; LIS3; LIS3 antibody; LOC106557476; M40; MGC117247; MGC16435; OK/SW cl0.56; OK/SWcl0.56; OK/SW-cl0.56; Talpha1 alpha-tubulin; TUBA1; Tuba-1; Tuba1a; TUBA1A antibody; TUBA3; TUBA3 antibody; TUBA6; TUBB1; TUBB5; tubulin alpha 1 A; tubulin alpha-1 chain; Tubulin alpha-1 A chain; tubulin alpha-1 A chain antibody; tubulin alpha-3 chain; tubulin alpha-3 chain antibody; Tubulin B-alpha-1; tubulin B-alpha-1 antibody; tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1 A; tubulin, alpha 6; tubulin, alpha, brain-specific; unnamed protein product
Common Name TUBA1A
Gene Symbol TUBA1A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LEHGIQPDGQMPSDKTIGGGDDSFNTFF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less