missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TTC7B (aa 564-652) Control Fragment Recombinant Protein

Catalog No. RP101939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101939 Supplier Invitrogen™ Supplier No. RP101939
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63750 (PA5-63750. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q86TV6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 145567
Name Human TTC7B (aa 564-652) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA408451; c14_5685; tetratricopeptide repeat domain 7 like 1; tetratricopeptide repeat domain 7 B; Tetratricopeptide repeat protein 7 B; tetratricopeptide repeat protein 7-like-1; TPR repeat protein 7 B; TPR repeat protein 7-like-1; Ttc7b; TTC7L1
Common Name TTC7B
Gene Symbol Ttc7b
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HYHDALNIIDMALSEYPENFILLFSKVKLQSLCRGPDEALLTCKHMLQIWKSCYNLTNPSDSGRGSSLLDRTIADRRQLNTITLPDFSD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less