missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 9530051K01Rik; E2IG4; E2-induced gene 4 protein; Early insulin-induced hepatic gene protein; EIIH; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; Lrrc54; Tsk; Tsku; tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushi, small leucine rich proteoglycan; Tsukushin; UNQ850/PRO1788 |
|---|---|
| Assay Range | 0.63 to 150 ng/mL |
| Assay Sensitivity | 0.63 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH474RBX5
|
Invitrogen™
EH474RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH474RB
|
Invitrogen™
EH474RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Tsukushi/TSK quantitates human Tsukushi/TSK in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Tsukushi/TSK.
The function of this protein remains unknown.Specifications
| 9530051K01Rik; E2IG4; E2-induced gene 4 protein; Early insulin-induced hepatic gene protein; EIIH; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; Lrrc54; Tsk; Tsku; tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushi, small leucine rich proteoglycan; Tsukushin; UNQ850/PRO1788 | |
| 0.63 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| TSKU | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| TSKU | |
| 4 hrs 45 mins |
| 0.63 to 150 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 25987 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |