missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSTD2 (aa 277-374) Control Fragment Recombinant Protein

Catalog No. RP92973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92973 Supplier Invitrogen™ Supplier No. RP92973
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55024 (PA5-55024. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TSTD2 is a protein coding gene.

Specifications

Accession Number Q5T7W7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 158427
Name Human TSTD2 (aa 277-374) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C9orf97; PP4189; rhodanese domain-containing protein 2; rhodanese like domain containing 2; thiosulfate sulfurtransferase (rhodanese)-like domain containing 2; thiosulfate sulfurtransferase like domain containing 2; thiosulfate sulfurtransferase -like domain containing 2; thiosulfate sulfurtransferase/rhodanese-like domain-containing protein 2; TSTD2
Common Name TSTD2
Gene Symbol TSTD2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KPGIHLSPGEFHKEVEKFLSQANQEQSDTILLDCRNFYESKIGRFQGCLAPDIRKFSYFPSYVDKNLELFREKRVLMYCTGGIRCERGSAYLKAKGVC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less