missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSSK4 (aa 238-315) Control Fragment Recombinant Protein

Catalog No. RP105156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105156 Supplier Invitrogen™ Supplier No. RP105156
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66343 (PA5-66343. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TSSK4 belongs to a family of serine/threonine kinases highly expressed in testis (Chen et al., 2005 [PubMed 15964553]).

Specifications

Accession Number Q6SA08
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 283629
Name Human TSSK4 (aa 238-315) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700020B19Rik; 4933424F08Rik; C14orf20; serine/threonine kinase 22 E; serine/threonine-protein kinase 22 E; STK22E; testis specific serine kinase 4; testis-specific kinase 4; testis-specific serine kinase 4; testis-specific serine/threonine protein kinase 5 variant alpha; testis-specific serine/threonine protein kinase 5 variant beta; testis-specific serine/threonine protein kinase 5 variant delta; testis-specific serine/threonine protein kinase 5 variant gamma; testis-specific serine/threonine-protein kinase 4; TSK4; TSK-4; Tssk4; TSSK-4; TSSK5
Common Name TSSK4
Gene Symbol TSSK4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PFDDTNLKKLLRETQKEVTFPANHTISQECKNLILQMLRQATKRATILDIIKDSWVLKFQPEQPTHEIRLLEAMCQLH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less