missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSPAN13 (aa 95-165) Control Fragment Recombinant Protein

Catalog No. RP88669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP88669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP88669 Supplier Invitrogen™ Supplier No. RP88669

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52434. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.

Specifications

Accession Number O95857
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 27075
Name Human TSPAN13 (aa 95-165) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1100001I23Rik; NET6; NET-6; tetraspan NET-6; Tetraspan Tspan-13; tetraspanin 13; tetraspanin-13; Tm4sf13; transmembrane 4 superfamily member 13; transmembrane 4 superfamily member tetraspan NET-6; Tspan13; tspan-13; UNQ260/PRO296
Common Name TSPAN13
Gene Symbol TSPAN13
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ACLALNQEQQGQLLEVGWNNTASARNDIQRNLNCCGFRSVNPNDTCLASCVKSDHSCSPCAPIIGEYAGEV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less