missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TRIM25 (NP_005073.2) VersaClone cDNA

Specifications
Specifications
| Gene | TRIM25 |
| Accession Number | NP_005073.2 |
| Gene ID (Entrez) | 7706 |
| Gene Alias | EC 6.3.2.19, EC 6.3.2.n3, EFP, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147, ZNF147tripartite motif protein TRIM25 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |