missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TOE1 (aa 437-509) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64401. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Inhibits cell growth rate and cell cycle. Induces CDKN1A expression as well as TGF-beta expression. Mediates the inhibitory growth effect of EGR1. Involved in the maturation of snRNAs and snRNA 3'-tail processing (PubMed:28092684). [UniProt].
Specifications
Specifications
| Accession Number | Q96GM8 |
| Concentration | 4.1 mg/mL |
| For Use With (Application) | Neutralization, Control |
| Formulation | PBS, 1M urea with no preservative; pH 7.4 |
| Gene ID (Entrez) | 114034 |
| Name | Human TOE1 (aa 437-509) Control Fragment |
| pH Range | 7.4 |
| Purification Method | Metal-chelate chromatography |
| Quantity | 100 μL |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |