missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM9B (aa 126-176) Control Fragment Recombinant Protein

Catalog No. RP96579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96579 Supplier Invitrogen™ Supplier No. RP96579
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58103 (PA5-58103. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TMEM9B, also termed as Transmembrane protein 9B encodes a 198 amino-acid protein that contains an N-terminal signal peptide, a single transmembrane region, three potential N-glycosylation sites, and three conserved cys-rich domains in the N-terminus. There is a dimer form of TMEM9B protein. TMEM9B mRNA is expressed in a wide range of tissues and cells. TMEM9B is a lysosomal transmembrane protein that regulates the activity of inflammatory signaling pathways.

Specifications

Accession Number Q9NQ34
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56674
Name Human TMEM9B (aa 126-176) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310004K06Rik; AW539847; C11orf15; D7H11orf15; ICRFP703B1614Q5.3; ICRFP703N2430Q5.3; RGD1310775; TMEM9 domain family member B; TMEM9 domain family, member B; Tmem9b; Transmembrane protein 9 B; UNQ712/PRO1375
Common Name TMEM9B
Gene Symbol TMEM9B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EPILKRRLFGHAQLIQSDDDIGDHQPFANAHDVLARSRSRANVLNKVEYAQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less