missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TMEM91 (aa 115-143) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100472
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62763 (PA5-62763. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function remains unknown.Specifications
| Q6ZNR0 | |
| Blocking Assay, Control | |
| 641649 | |
| 100 μL | |
| dispanin subfamily C member 3; DSPC3; IFITMD6; interferon induced transmembrane protein domain containing 6; TMEM91; Transmembrane protein 91 | |
| TMEM91 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human TMEM91 (aa 115-143) Control Fragment | |
| RUO | |
| TMEM91 | |
| Unconjugated | |
| Recombinant | |
| FCLAQKTNKAWAKGDIQGAGAASRRAFLL | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |