missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM155 Control Fragment Recombinant Protein

Catalog No. RP108687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108687 Supplier Invitrogen™ Supplier No. RP108687

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (30%), Rat (30%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-85038 (PA5-85038. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TMEM155 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.

Specifications

Accession Number Q4W5P6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 132332
Name Human TMEM155 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias FLJ30834; protein TMEM155; TMEM155; transmembrane protein 155
Common Name TMEM155
Gene Symbol TMEM155
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AVDAELMPSGAILQNKRENLPRVCHALAFLGMARCQDLFLVRLQGWKLGTR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less