missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM136 (aa 19-42) Control Fragment Recombinant Protein

Catalog No. RP107997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107997 Supplier Invitrogen™ Supplier No. RP107997

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67346. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TMEM136 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.

Specifications

Accession Number Q6ZRR5
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 219902
Name Human TMEM136 (aa 19-42) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AA396355; Gm184; RGD1565079; TLC domain containing 5; TLC domain-containing protein 5; TLC domain-containing protein 5; transmembrane protein 136; TLCD5; TMEM136; Transmembrane protein 136
Common Name TMEM136
Gene Symbol TMEM136
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LYISFCHLNKHRSYEWSCRLVTFT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less