missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TM4SF18 (aa 32-51) Control Fragment Recombinant Protein

Catalog No. RP107690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107690 Supplier Invitrogen™ Supplier No. RP107690

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67048 (PA5-67048. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TM4SF18 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.

Specifications

Accession Number Q96CE8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 116441
Name Human TM4SF18 (aa 32-51) Control Fragment
Quantity 100 μL
Gene Alias AAB5; L6D; TM4SF18; transmembrane 4 L six family member 18; transmembrane 4 L6 family member 18
Common Name TM4SF18
Gene Symbol TM4SF18
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NGQTSYASSNKLTNYVWYFE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less