missing translation for 'onlineSavingsMsg'
Learn More

Human TLR9 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Catalog No. IC3658N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3658N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC3658N100 Supplier Bio-Techne Supplier No. IC3658N100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen TLR9
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 229106
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_059138
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Gene Symbols TLR9
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TLR9 Accession # NP_059138
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54106
Test Specificity Detects human TLR9. Stains human TLR9-transfected cells but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgM
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)