missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIMP-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Description
TIMP-1 Monoclonal specifically detects TIMP-1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | TIMP-1 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 63515 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P01033 |
| Gene Alias | CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 |
| Show More |
For Research Use Only (RUO)