missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TGF-beta RIII ELISA Kit
Description
Human TGF-beta RIII quantitates human TGF-beta RIII in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TGF-beta RIII.
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
Specifications
| Accession Number | Q03167 |
| Assay Range | 40-10000 pg/mL |
| Assay Sensitivity | 40 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | 1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III; |
| Show More |