missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGF beta 3 Carrier-Free, Recombinant Protein, eBioscience™
Recombinant Protein
$8500.00 - $8500.00
Specifications
| Accession Number | P10600 |
|---|---|
| Formulation | protein with 0.1% TFA and no preservative |
| Gene ID (Entrez) | 7043 |
| Molecular Weight (g/mol) | 25.0kDa |
| Name | Human TGF beta 3 Carrier-Free |
Description
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity. Description: Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity. The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity. Applications Reported: Recombinant human TGF beta 3 is biologically active and can inhibit IL-4-induced proliferation of mouse HT-2 cells. Applications Tested: The ED50 of this protein, as measured by inhibition of IL-4-induced proliferation of mouse HT-2 cells, is less than or equal to 0.05ng/mL, which corresponds to a specific activity of greater than 2 x 10^7 Units/mg. Source: E. coli (amino acids Ala301-Ser412; accession # NP_003230.1). Bioactivity: The ED50 of this protein, as measured by inhibition of IL-4-induced proliferation of mouse HT-2 cells, is less than or equal to 0.05ng/mL, which corresponds to a specific activity of greater than 2 x 10e7 Units/mg. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE and HPLC. Molecular Weight: 25.0 kDa. Storage and handling: For best recovery, quick spin vial prior to opening. Reconstitute in 5-10 mM citric acid to a concentration of 0.1-1.0mg/mL. Do not vortex. Use in sterile environment. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier-protein (e.g., 0.1% BSA) at less than or equal to -20°C.Specifications
| P10600 | |
| 7043 | |
| Human TGF beta 3 Carrier-Free | |
| E. coli | |
| RUO | |
| Less than 0.1ng/μg cytokine as determined by the LAL assay | |
| TGFB3 | |
| Recombinant Protein, Carrier-Free | |
| Unconjugated | |
| E. coli | |
| Lyophilized | |
| Affinity chromatography |
| protein with 0.1% TFA and no preservative | |
| 25.0kDa | |
| SDS-PAGE and HPLC | |
| -20°C | |
| E. coli (amino acids Ala 301 - Ser412; Accession # NP_003230.1) | |
| Human TGF beta 3 Carrier-Free | |
| The ED50 of this protein, as measured by inhibition of IL-4-induced proliferation of mouse HT-2 cells, is less than or equal to 0.05ng/mL, which corresponds to a specific activity of greater than 2 x 107 Units/mg. | |
| Human | |
| Hu | |
| Recombinant | |
| Recombinant |