missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TEX101 (aa 25-99) Control Fragment Recombinant Protein

Catalog No. RP99089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99089 Supplier Invitrogen™ Supplier No. RP99089

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (49%), Rat (49%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59784. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May play a role in signal transduction and promote protein tyrosine phosphorylation. Target protein is a glycoprotein whose aggregation induced a rapid increase in tyrosine phosphorylation of Syk kinase and LAT adaptor, calcium flux, and release of secretory components.

Specifications

Accession Number Q9BY14
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 83639
Name Human TEX101 (aa 25-99) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias cancer/testis antigen 131; cell surface receptor NYD-SP8; CT131; GTPR867; NYD-SP8; PRO1884; Scleroderma-associated autoantigen; SGRG; SPATA44; spermatogenesis associated 44; spermatogenesis-related gene protein; TES101RP; testis expressed 101; testis expressed sequence 101; testis-expressed protein 101; testis-expressed sequence 101 protein; testis-specific protein TES101RP; TEX101; UNQ867/PRO1884
Common Name TEX101
Gene Symbol TEX101
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ELYCQKGLSMTVEADPANMFNWTTEEVETCDKGALCQETILIIKAGTETAILATKGCIPEGEEAITIVQHSSPPG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less