missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TCF9 Control Fragment Recombinant Protein

Catalog No. RP108129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108129 Supplier Invitrogen™ Supplier No. RP108129

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67476 (PA5-67476. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.

Specifications

Accession Number P16383
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6936
Name Human TCF9 Control Fragment
Quantity 100 μL
Gene Alias A130099G21; AW146020; C2orf3; DNABF; GC binding factor; GC bindng factor; GCF; GCF2; GCFC2; GC-rich sequence DNA binding factor 2; GC-rich sequence DNA-binding factor; GC-rich sequence DNA-binding factor 2; RGD1304792; TCF9; TCF-9; Transcription factor 9; transcription factor 9 (binds GC-rich sequences)
Common Name TCF9
Gene Symbol GCFC2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ESEPDDHEKRIPFTLRPQTLRQRMAEESISRNEETSEESQEDEKQDTWEQQQMRKAVKIIEERDIDLSCGSGSSKVKKFDTSISFP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less