missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TBCC (aa 93-168) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP94798

Catalog No. RP94798


Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (64%), Rat (64%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57143 (PA5-57143. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Tubulin-folding protein; involved in the final step of the tubulin folding pathway.
Specifications

Specifications

Q15814
Blocking Assay, Control
6903
100 μL
2810055C19Rik; CFC; EGK_14912; Tbcc; tubulin folding cofactor C; tubulin-folding cofactor C; Tubulin-specific chaperone C
TBCC
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human TBCC (aa 93-168) Control Fragment
RUO
TBCC
Unconjugated
Recombinant
GLQKLINDSVFFLAAYDLRQGQEALARLQAALAERRRGLQPKKRFAFKTRGKDAASSTKVDAAPGIPPAVESIQDS
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions