missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TAF1L (aa 1807-1826) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100035
Description
Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63325 (PA5-63325. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TAF1L, also named as TAF (II)210, belongs to the TAF1 family. It may act as a functional substitute for TAF1/TAFII250 during male meiosis, when sex chromosomes are transcriptionally silenced.Specifications
| Q8IZX4 | |
| Blocking Assay, Control | |
| 138474 | |
| 100 μL | |
| TAF(II)210; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 210 kDa-like; TAF1L; TAF2A2; TATA-box binding protein associated factor 1 like; TBP-associated factor 1-like; TBP-associated factor 210 kDa; TBP-associated factor RNA polymerase 1-like; transcription initiation factor TFIID 210 kDa subunit; Transcription initiation factor TFIID subunit 1-like | |
| TAF1L | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human TAF1L (aa 1807-1826) Control Fragment | |
| RUO | |
| TAF1L | |
| Unconjugated | |
| Recombinant | |
| PKQPFMLQHASGEHKDGHGK | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |