missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human t-Plasminogen Activator (NP_000921.1) VersaClone cDNA
GREENER_CHOICE

Catalog No. RDC3776
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC3776 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDC3776 Supplier R&D Systems™ Supplier No. FAB10988G100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Gene t-Plasminogen Activator/tPA
Accession Number NP_000921.1
Gene ID (Entrez) 5327
Gene Alias Alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, PLAT, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, TPA, T-PA, t-plasminogen activator, tPlasminogen Activator
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg