missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SYTL1 (aa 43-131) Control Fragment Recombinant Protein

Catalog No. RP94260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94260 Supplier Invitrogen™ Supplier No. RP94260

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58126 (PA5-58126. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SYTL1 may play a role in vesicle trafficking. Binds phosphatidylinositol 3,4,5-trisphosphate. Acts as a RAB27A effector protein and may play a role in cytotoxic granule exocytosis in lymphocytes.

Specifications

Accession Number Q8IYJ3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84958
Name Human SYTL1 (aa 43-131) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias exophilin-7; FLJ14996; JFC1; NADPH oxidase-related, C2 domain-containing protein; Protein JFC1; Protein SLP1; SB146; SLP1; synaptotagmin like 1; synaptotagmin-like 1; synaptotagmin-like protein 1; SYTL1
Common Name SYTL1
Gene Symbol SYTL1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EAIAGVLQRDARLRQLEEGRVSKLRASVADPGQLKILTGDWFQEARSQRHHNAHFGSDLVRASMRRKKSTRGDQAPGHDREAEAAVKEK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less