missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SYT15 (aa 209-273) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP105645
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66691 (PA5-66691. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in the trafficking and exocytosis of secretory vesicles in non-neuronal tissues.Specifications
| Q9BQS2 | |
| Blocking Assay, Control | |
| 83849 | |
| 100 μL | |
| chr10 synaptotagmin; CHR10SYT; synaptotagmin 15; synaptotagmin XV; synaptotagmin XV-a; synaptotagmin-15; SYT15; SytXV | |
| SYT15 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human SYT15 (aa 209-273) Control Fragment | |
| RUO | |
| SYT15 | |
| Unconjugated | |
| Recombinant | |
| QFDEHFIFQVSSKTITQRVLKFSVYHVDRQRKHQLLGQVLFPLKNETLVGDCRRVIWRDLEAESL | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |