missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Syndecan 3 (aa 65-167) Control Fragment Recombinant Protein

Catalog No. RP104845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104845 Supplier Invitrogen™ Supplier No. RP104845
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65494 (PA5-65494. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene belongs to the syndecan proteoglycan family. It may play a role in the organization of cell shape by affecting the actin cytoskeleton, possibly by transferring signals from the cell surface in a sugar-dependent mechanism. Allelic variants of this gene have been associated with obesity.

Specifications

Accession Number O75056
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9672
Name Human Syndecan 3 (aa 65-167) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Kiaa0468; mKIAA0468; Neuroglycan; N-syndecan; SDC3; SDCN; syn-3; Synd3; syndecan 1; syndecan 3; syndecan neural type; syndecan proteoglycan 3; syndecan-3; wu:fa97a01
Common Name Syndecan 3
Gene Symbol Sdc3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GDDDSFPDDELDDLYSGSGSGYFEQESGIETAMRFSPDVALAVSTTPAVLPTTNIQPVGTPFEELPSERPTLEPATSPLVVTEVPEEPSQRATTVSTTMATTA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less